Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017944.1 | NP_001017944.1 | 2 - 3 | 265 | 89 | |
GenBank mRNA | BC112631.1 | - | 2 - 3 | 285 | 89 | |
BT021177.1 | - | 2 - 3 | 265 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |