Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024494.1 | NP_001019665.1 | 7 - 8 | 1489 | 72 | |
GenBank mRNA | BC148055.1 | - | 7 - 8 | 1486 | 72 | |
BT020770.1 | - | 7 - 8 | 1489 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |