Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034690.2 | NP_001029862.1 | 3 - 4 | 323 | 81 | |
GenBank mRNA | BC102049.1 | - | 3 - 4 | 324 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |