Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035450.1 | NP_001030527.1 | 5 - 6 | 493 | 106 | |
GenBank mRNA | BC119872.1 | - | 5 - 6 | 528 | 106 | |
BT020837.1 | - | 5 - 6 | 493 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |