Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075320.1 | NP_001068788.1 | 2 - 3 | 415 | 88 | |
GenBank mRNA | BT020787.1 | - | 2 - 3 | 415 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |