Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076970.1 | NP_001070438.1 | 7 - 8 | 758 | 114 | |
GenBank mRNA | BC120025.1 | - | 7 - 8 | 762 | 114 | |
BT021195.1 | - | 7 - 8 | 758 | 114 | ||
BT021606.1 | - | 7 - 8 | 754 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |