Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5PF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174717.3 | NP_777142.1 | 2 - 3 | 213 | 81 | |
GenBank mRNA | BC103429.1 | - | 2 - 3 | 216 | 81 | |
M19217.1 | - | 2 - 3 | 216 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |