Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034265.2 | NP_001029437.1 | 4 - 5 | 623 | 102 | |
GenBank mRNA | BC102592.1 | - | 4 - 5 | 625 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |