Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034299.1 | NP_001029471.1 | 5 - 6 | 639 | 110 | |
GenBank mRNA | BC102308.1 | - | 5 - 6 | 639 | 110 | |
BT021601.1 | - | 5 - 6 | 569 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |