Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETHE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034344.1 | NP_001029516.1 | 3 - 4 | 420 | 113 | |
GenBank mRNA | BC102496.1 | - | 3 - 4 | 420 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |