Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034622.3 | NP_001029794.1 | 2 - 3 | 491 | 75 | |
GenBank mRNA | BC105211.1 | - | 2 - 3 | 227 | 75 | |
EE227096.1 | - | 2 - 3 | 491 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |