Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035305.2 | NP_001030382.1 | 10 - 11 | 1708 | 123 | |
GenBank mRNA | BC107540.1 | - | 10 - 11 | 1709 | 123 | |
BT030747.1 | - | 10 - 11 | 1601 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |