Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPH3AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035494.1 | NP_001030571.1 | 8 - 9 | 933 | 126 | |
GenBank mRNA | BC102095.1 | - | 8 - 9 | 933 | 126 | |
BT021718.1 | - | 8 - 9 | 974 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |