Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076329.2 | NP_001069797.1 | 134 | 63 | ||
GenBank mRNA | BC108233.1 | - | 136 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus |