Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077077.2 | NP_001070545.1 | 1 - 2 | 300 | 121 | |
GenBank mRNA | BC111617.1 | - | 1 - 2 | 301 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |