Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014930.3 | NP_001014930.1 | 1 - 2 | 202 | 77 | |
GenBank mRNA | BC109791.1 | - | 1 - 2 | 203 | 77 | |
BT021045.1 | - | 1 - 2 | 97 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |