Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037625.2 | NP_001032714.1 | 2 - 3 | 687 | 141 | |
GenBank mRNA | BC109601.1 | - | 2 - 3 | 688 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |