Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038034.2 | NP_001033123.1 | 6 - 7 | 553 | 146 | |
GenBank mRNA | BC109637.1 | - | 6 - 7 | 554 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |