Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038044.2 | NP_001033133.1 | 8 - 9 | 1068 | 76 | |
GenBank mRNA | BC111283.1 | - | 8 - 9 | 1069 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |