Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038192.2 | NP_001033281.1 | 1 - 2 | 269 | 86 | |
GenBank mRNA | BC109681.1 | - | 1 - 2 | 270 | 86 | |
BT030668.1 | - | 1 - 2 | 239 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |