Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038219.2 | NP_001033308.2 | 1 - 2 | 91 | 89 | |
GenBank mRNA | BC109498.1 | - | 1 - 2 | 92 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |