Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045880.2 | NP_001039345.1 | 6 - 7 | 711 | 87 | |
GenBank mRNA | AY630919.1 | - | 1 - 2 | 251 | 87 | |
BC110146.1 | - | 6 - 7 | 712 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |