Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046186.2 | NP_001039651.1 | 3 - 4 | 711 | 86 | |
GenBank mRNA | BC112464.1 | - | 3 - 4 | 712 | 86 | |
BT029911.1 | - | 4 - 5 | 734 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |