Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046387.1 | NP_001039852.1 | 5 - 6 | 981 | 105 | |
GenBank mRNA | BC111154.1 | - | 5 - 6 | 981 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |