Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046407.1 | NP_001039872.1 | 1 - 2 | 365 | 145 | |
GenBank mRNA | BC110223.1 | - | 1 - 2 | 365 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |