Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205479.1 | NP_001192408.1 | 2 - 3 | 592 | 82 | |
GenBank mRNA | BC112863.1 | - | 2 - 3 | 544 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |