Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046529.2 | NP_001039994.1 | 6 - 7 | 757 | 133 | |
GenBank mRNA | BC112495.1 | - | 6 - 7 | 758 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |