Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076507.2 | NP_001069975.1 | 2 - 3 | 546 | 74 | |
GenBank mRNA | BC109514.1 | - | 2 - 3 | 547 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |