Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077123.2 | NP_001070591.1 | 3 - 4 | 239 | 69 | |
GenBank mRNA | BC110123.1 | - | 3 - 4 | 240 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |