Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079630.1 | NP_001073098.1 | 1 - 2 | 384 | 85 | |
GenBank mRNA | BC109649.1 | - | 1 - 2 | 384 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |