Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPADH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174616.3 | NP_777041.1 | 1 - 2 | 535 | 106 | |
GenBank mRNA | BC110191.1 | - | 1 - 2 | 535 | 106 | |
M84603.1 | - | 1 - 2 | 54 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |