Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046118.2 | NP_001039583.1 | 2 - 3 | 248 | 68 | |
GenBank mRNA | BC105438.1 | - | 2 - 3 | 250 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |