Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046170.1 | NP_001039635.1 | 11 - 12 | 1643 | 108 | |
GenBank mRNA | BC112461.1 | - | 11 - 12 | 1643 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |