Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046336.1 | NP_001039801.1 | 10 - 11 | 1473 | 70 | |
GenBank mRNA | BC114078.1 | - | 10 - 11 | 1473 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |