Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046474.2 | NP_001039939.1 | 4 - 5 | 933 | 82 | |
GenBank mRNA | AJ871967.1 | - | 4 - 5 | 907 | 82 | |
AJ871968.1 | - | 4 - 5 | 763 | 82 | ||
BC118079.1 | - | 4 - 5 | 934 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |