Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075173.1 | NP_001068641.1 | 6 - 7 | 829 | 69 | |
GenBank mRNA | BC112659.1 | - | 6 - 7 | 829 | 69 | |
DQ087595.1 | - | 2 - 3 | 290 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |