Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075363.2 | NP_001068831.1 | 3 - 4 | 220 | 88 | |
GenBank mRNA | BC118342.1 | - | 3 - 4 | 221 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |