Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075744.1 | NP_001069212.1 | 4 - 5 | 638 | 106 | |
GenBank mRNA | BC118205.1 | - | 4 - 5 | 638 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |