Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076261.1 | NP_001069729.1 | 5 - 6 | 846 | 88 | |
GenBank mRNA | BC133446.1 | - | 5 - 6 | 776 | 88 | |
BT025400.1 | - | 5 - 6 | 846 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |