Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076275.1 | NP_001069743.1 | 2 - 3 | 268 | 74 | |
GenBank mRNA | BC116058.1 | - | 2 - 3 | 268 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |