Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075162.2 | NP_001068630.1 | 1 - 2 | 120 | 66 | |
NM_001163274.3 | NP_001156746.2 | 1 - 2 | 120 | 66 | ||
GenBank mRNA | BC120448.1 | - | 1 - 2 | 40 | 66 | |
BE751148.1 | - | 1 - 2 | 120 | 66 | ||
EH159834.1 | - | 1 - 2 | 86 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |