Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075266.1 | NP_001068734.1 | 3 - 4 | 375 | 85 | |
GenBank mRNA | BC118195.1 | - | 3 - 4 | 375 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |