Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075419.1 | NP_001068887.1 | 2 - 3 | 121 | 83 | |
GenBank mRNA | BC120342.1 | - | 2 - 3 | 121 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |