Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075431.2 | NP_001068899.1 | 4 - 5 | 465 | 125 | |
GenBank mRNA | BC120124.1 | - | 4 - 5 | 466 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |