Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075603.1 | NP_001069071.1 | 5 - 6 | 595 | 126 | |
GenBank mRNA | BC120016.1 | - | 5 - 6 | 595 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |