Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076825.1 | NP_001070293.1 | 2 - 3 | 154 | 57 | |
GenBank mRNA | BC123759.1 | - | 2 - 3 | 154 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |