Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015635.2 | NP_001015635.1 | 2 - 3 | 382 | 80 | |
GenBank mRNA | BC142345.1 | - | 2 - 3 | 395 | 80 | |
BT020927.1 | - | 2 - 3 | 314 | 80 | ||
BT029844.1 | - | 2 - 3 | 382 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |