Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083498.1 | NP_001076967.1 | 4 - 5 | 515 | 62 | |
GenBank mRNA | BC123637.1 | - | 4 - 5 | 515 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |