Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111325.1 | NP_001104795.1 | 1 - 2 | 491 | 96 | |
GenBank mRNA | AB110600.1 | - | 1 - 2 | 96 | ||
AB257560.1 | - | 1 - 2 | 96 | |||
AF091714.1 | - | 1 - 2 | 96 | |||
BC118336.1 | - | 1 - 2 | 491 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |