Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174522.3 | NP_776947.1 | 2 - 3 | 279 | 64 | |
GenBank mRNA | BC103117.1 | - | 2 - 3 | 251 | 64 | |
DV846222.1 | - | 280 | 64 | |||
X56857.1 | - | 226 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |